missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Platinum(IV) oxide hydrate, typical Pt-content 79-84%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 211920250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Platinum(IV) oxide hydrate | |
25g | |
PtO2 | |
1479 | |
14, 7527 | |
Solubility in water: insoluble. Other solubilities: soluble in conc. acids and dil.sol. of koh | |
227.09 | |
Glass bottle |
52785-06-5 | |
O2Pt·xH2O | |
MFCD00066964 | |
01,890; 15,268 | |
Adams' catalyst | |
227.09 | |
typical Pt-content 79-84% |
Safety and Handling
GHS H Statement
May cause fire or explosion; strong oxidizer.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major
GHS Signal Word: Danger
RUO â Research Use Only