Please login to your online account to display your discounted pricing

Platinum(IV) oxide hydrate, typical Pt-content 79-84%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  211920010

 View more versions of this product

Catalog No. AC211920010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 227.09
Quantity 1g
Color Brown to black
Merck Index 14, 7527
Assay 79 to 84% (Pt)
UN Number 1479
Synonym Adams' catalyst
Chemical Name or Material Platinum(IV) oxide hydrate
Surface Area 90 m2/g min. (BET)
MDL Number MFCD00066964
CAS 52785-06-5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May intensify fire; oxidizer.
Packaging Glass bottle
Molecular Formula O2Pt·xH2O
Linear Formula PtO2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Platinum(IV) oxide hydrate
Assay 79 to 84% (Pt)
Molecular Formula O2Pt·xH2O
Linear Formula PtO2
Formula Weight 227.09
MDL Number MFCD00066964
Synonym Adams' catalyst
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May intensify fire; oxidizer.

SDS & Safety Documents