Learn More
Platinum(IV) oxide, 83% Pt, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 195320010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum(IV) oxide | |
83% Pt | |
Brown | |
1 g | |
PtO2 | |
1479 | |
Adams' catalyst | |
YKIOKAURTKXMSB-UHFFFAOYSA-N | |
dioxoplatinum | |
227.09 | |
Glass bottle |
450.0°C | |
1314-15-4 | |
Powder | |
O2Pt | |
MFCD00011184 | |
15, 7642 | |
Solubility in water: insoluble | |
O=[Pt]=O | |
227.08 | |
83% (Pt) |
Chemical Identifiers
1314-15-4 | |
227.08 | |
YKIOKAURTKXMSB-UHFFFAOYSA-N | |
dioxoplatinum |
O2Pt | |
MFCD00011184 | |
Adams' catalyst | |
O=[Pt]=O |
Safety and Handling
GHS H Statement
May cause fire or explosion; strong oxidizer.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major
GHS Signal Word: Danger
EINECSNumber : 215-223-0
TSCA : TSCA
RUO – Research Use Only