Please login to your online account to display your discounted pricing

Platinum(IV) oxide, 83% Pt, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC195320050

 View more versions of this product

Catalog No. AC195320050

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Platinum(IV) oxide
Name Note 83% Pt
CAS 1314-15-4
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye damage.
May intensify fire; oxidizer.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 215-223-0
TSCA TSCA
Physical Form Powder
Assay ≥79% (Pt)
Molecular Formula O2Pt
Linear Formula PtO2
Formula Weight 227.09
MDL Number MFCD00011184
UN Number 1479
Merck Index 15, 7642
Synonym Adams' catalyst
Packaging Glass bottle
Color Brown
Quantity 5g

Specifications

Chemical Name or Material Platinum(IV) oxide
Name Note 83% Pt
CAS 1314-15-4
Assay ≥79% (Pt)
Molecular Formula O2Pt
Linear Formula PtO2
Formula Weight 227.09
MDL Number MFCD00011184
Synonym Adams' catalyst
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye damage.
May intensify fire; oxidizer.

SDS & Safety Documents