Please login to your online account to display your discounted pricing

Platinum(IV) iodide, 99.95% (Metals basis), Pt 27.3% min., Alfa Aesar™, Premion™

Chemical Identifiers

Chemical Name or Material Platinum(IV) iodide
CAS 7790-46-7
Physical Form Powder
Assay Percent Range 99.95% (metals basis)
Molecular Formula I4Pt
Formula Weight 702.7
MDL Number MFCD00016292
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4040101

 
alfa aesar
4040101
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4040103

 
alfa aesar
4040103
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4040106

 
alfa aesar
4040106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 702.7
Quantity 0.25g
Color Black
DOT Information DOT Class: 8, Packaging Group: III
Chemical Name or Material Platinum(IV) iodide
EINECS Number 232-207-9
Assay Percent Range 99.95% (metals basis)
Molecular Formula I4Pt
Appearance Black powder
MDL Number MFCD00016292
CAS 7790-46-7
Health Hazard 1 H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.
Health Hazard 2 When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Composition Pt 27.3% min.
Safety and Handling

H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).