Please login to your online account to display your discounted pricing

Platinum(IV) chloride, 99%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC195402500

 View more versions of this product

Catalog No. AC195402500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 336.89
Quantity 250mg
Color Brown
EINECS Number 236-645-1
TSCA TSCA
Assay 57.0 to 58.5% (Pt)
RTECS Number TP2275500
Chemical Name or Material Platinum(IV) chloride
Iron (Fe) 50ppm max.
Lead (Pb) 25ppm max.
Magnesium (Mg) 250ppm max.
Name Note 99%
Packaging Glass bottle
Silicon (Si) 200ppm max.
Assay Percent Range 99%
Molecular Formula Cl4Pt
Linear Formula PtCl4
MDL Number MFCD00011182
CAS 13454-96-1
Health Hazard 1 GHS Signal Word: Danger
Additional Information Ru+Rh+Ir+Pd <500ppm
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Toxic if swallowed.
Calcium (Ca) 250ppm max.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Copper (Cu) 25ppm max.

Specifications

Chemical Name or Material Platinum(IV) chloride
Name Note 99%
CAS 13454-96-1
Assay 57.0 to 58.5% (Pt)
Assay Percent Range 99%
Molecular Formula Cl4Pt
Linear Formula PtCl4
Formula Weight 336.89
MDL Number MFCD00011182
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Toxic if swallowed.

SDS & Safety Documents