Learn More
Platinum(IV) chloride, 99.9% (metals basis), Pt 57% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01104503
Description
Platinum(IV) chloride is used as a catalyst and analytical reagent. Further, it is used in the production of semiconductor epitaxy and in electroplating process. It is involved in the preparation of platinum(II) chloride .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum(IV) chloride | |
Pt 57% min. | |
Red to Brown | |
1 g | |
Cl4Pt | |
platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4 | |
FBEIPJNQGITEBL-UHFFFAOYSA-J | |
tetrachloroplatinum | |
26031 | |
Hygroscopic |
370°C (decomposition) | |
13454-96-1 | |
Crystalline | |
99.9% (metals basis) | |
MFCD00011182 | |
Soluble in water and acetone. Slightly soluble in ethanol and insoluble in ether. | |
Cl[Pt](Cl)(Cl)Cl | |
336.88 | |
336.9 | |
4.303 |
Chemical Identifiers
13454-96-1 | |
336.88 | |
FBEIPJNQGITEBL-UHFFFAOYSA-J | |
26031 | |
Cl[Pt](Cl)(Cl)Cl |
Cl4Pt | |
MFCD00011182 | |
platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4 | |
tetrachloroplatinum |
Safety and Handling
P260-P264b-P270-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H301-H314-H317-H334
DOTInformation : DOT Class: 8; Packaging Group: II
EINECSNumber : 236-645-1
RTECSNumber : TP2275500
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only