Alfa Aesar™ Platinum Iridium wire, 1.0mm (0.04 in.) dia., hard
Manufacturer:  Alfa Aesar™ 44732BS
 View more versions of this product
Catalog No. AA44732BS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Pt:Ir; 80:20 wt%
≈0.85g/5cm
1.0mm (0.04 in.) dia., Hard
Odorless
Platinum Iridium wire
10cm
MFCD02091683
Wire
Documents