Learn More
Platinum Iridium wire, 0.2mm (0.008in) dia, annealed, 99.9+% (metals basis), Thermo Scientific Chemicals
Form: Approx. 0.7g/10cm; Temper: soft; Formula: Pt:Ir; 80:20 wt%
$161.92 - $809.60
Chemical Identifiers
CAS | 7440-06-4 |
---|---|
Molecular Formula | Pt |
Molecular Weight (g/mol) | 195.08 |
MDL Number | MFCD00011179 |
InChI Key | BASFCYQUMIYNBI-UHFFFAOYSA-N |
Synonym | black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion |
PubChem CID | 23939 |
ChEBI | CHEBI:33400 |
IUPAC Name | platinum |
SMILES | [Pt] |
Description
Platinum-iridium wire is used in platinum jewellery, particularly watch cases. They have been used for spinnerets in the manufacture of synthetic fibres, in metrology and alloy is fabrication of metal microelectrodes. It is used in electrical stimulation of nervous tissue and electrophysiological recordings. Pt/Ir alloy has been used in mechanical and electrochemical devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-06-4 | |
195.08 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
23939 | |
platinum |
Pt | |
MFCD00011179 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
CHEBI:33400 | |
[Pt] |
Specifications
Silver to Gray | |
Silver gray wire | |
Pt | |
10 cm | |
Insoluble in water and acid. | |
[Pt] | |
195.08 | |
CHEBI:33400 | |
Wire |
7440-06-4 | |
>99.9% | |
MFCD00011179 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
platinum | |
23939 | |
Odorless | |
Platinum Iridium |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only