missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum Iridium Wire, 0.25mm (0.01 in.) dia., Alfa Aesar™

Form: Approx. 0.27g/25cm, Temper: soft

Manufacturer:  Alfa Aesar 43275BY

 View more versions of this product

Catalog No. AA43275BY



Specifications

Specifications

Platinum Iridium
100cm
7440-06-4
Pt
black
[Pt]
195.08
CHEBI:33400
Silver to Gray
Silver gray wire
Odorless
MFCD02091683
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Wire, 0.25mm (0.01 in.) dia.

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD02091683
black
CHEBI:33400
[Pt]
Documents