missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum(II) iodide, Premion™, 99.99% (metals basis), Pt 43.0% min

Manufacturer:  Alfa Aesar™ 1217001

 View more versions of this product

Catalog No. AA1217001



Specifications

Specifications

Platinum(II) iodide
(metals basis)
platinum ii iodide, platinum iodide pti2, platinum ii iodide, platinum ii iodide, premion, platinum ii iodide,premion metalsbasis ,ptmin
448.9
99.99%
Pt 43.0% min.
325°C (decomposition)
7790-39-8
MFCD00011183
Insoluble in water, alkali iodides
Powder
Premion™
Odorless
0.25g

Chemical Identifiers

7790-39-8
MFCD00011183
platinum ii iodide, platinum iodide pti2, platinum ii iodide, platinum ii iodide, premion, platinum ii iodide,premion metalsbasis ,ptmin
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents