Learn More
Platinum(II) iodide, Premion™, 99.99% (metals basis), Pt 43.0% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01217003
Description
Platinum(II) iodide is used in pharmaceuticals and medical applications. It is also used to form cis and trans complexes with propan-1-amine. It is utilized in the catalytic hydration reaction of alkynes such as 2-hexyne, 4-methyl-2-pentyne, and 4,4dimethyl-2- pentyne . It is also used in the preparation of diiodo(2,9-dimethyl-1,10-phenanthroline)platinum(II).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum(II) iodide | |
7790-39-8 | |
1 g | |
I2Pt | |
platinum ii iodide, platinum iodide pti2, platinum ii iodide, premion, platinum ii iodide,premion metalsbasis ,ptmin | |
ZXDJCKVQKCNWEI-UHFFFAOYSA-L | |
platinum(2+) diiodide | |
448.9 | |
Premion™ | |
Odorless |
325°C (decomposition) | |
Powder | |
(metals basis) | |
MFCD00011183 | |
Insoluble in water,alkali iodides. | |
[I-].[I-].[Pt++] | |
448.89 | |
99.99% | |
Pt 43.0% min. |
Chemical Identifiers
7790-39-8 | |
448.89 | |
ZXDJCKVQKCNWEI-UHFFFAOYSA-L | |
platinum(2+) diiodide |
I2Pt | |
MFCD00011183 | |
platinum ii iodide, platinum iodide pti2, platinum ii iodide, premion, platinum ii iodide,premion metalsbasis ,ptmin | |
[I-].[I-].[Pt++] |
Safety and Handling
GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 232-204-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only