Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum(II) cyanide, 98+%

Chemical Identifiers

Chemical Name or Material Platinum(II) cyanide
CAS 592-06-3
Molecular Formula C2N2Pt
Linear Formula Pt(CN)2
Formula Weight 247.13
MDL Number MFCD00016288
UN Number UN1588
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2305502

 
alfa aesar™
B2305502
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2305588

 
alfa aesar™
B2305588
2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 247.13
Chemical Name or Material Platinum(II) cyanide
Assay ≥98%
Molecular Formula C2N2Pt
Linear Formula Pt(CN)2
MDL Number MFCD00016288
CAS 592-06-3
UN Number UN1588
Quantity 0.5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CYANIDES, INORGANIC, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.