Learn More
Platinum(II) chloride, 99.9% (metals basis), Pt 73% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01104406
Description
Platinum(II) chloride is used as a catalyst for a variety of C-C bond, C-O bond and C-N bond formation reactions. It acts as a precursor to prepare other platinum compounds such as cisplatin and dichloro(cycloocta-1,5-diene)platinum(II). It is a well known chemotherapy drug.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum(II) chloride | |
10025-65-7 | |
Crystalline | |
(metals basis) | |
PtCl2 | |
UN3261 | |
Soluble in ammonia and hydrochloric acid; Insoluble in water,alcohol and ether. | |
[Cl-].[Cl-].[Pt++] | |
265.98 | |
99.9% | |
5.87 g/mL |
581°C (decomposition) | |
Brown-Green | |
5 g | |
Cl2Pt | |
MFCD00011181 | |
14,7528 | |
CLSUSRZJUQMOHH-UHFFFAOYSA-L | |
platinum(2+) dichloride | |
266 | |
Pt 73% min. |
Chemical Identifiers
10025-65-7 | |
265.98 | |
CLSUSRZJUQMOHH-UHFFFAOYSA-L | |
[Cl-].[Cl-].[Pt++] |
Cl2Pt | |
MFCD00011181 | |
platinum(2+) dichloride |
Safety and Handling
GHS H Statement
H334-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
P260-P264b-P272-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
H314-H317-H334
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 233-034-1
RTECSNumber : TP2275000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only