missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum(II) bromide, 99.9% (metals basis), Pt 54.5% min

Manufacturer:  Alfa Aesar™ 1217203

 View more versions of this product

Catalog No. AA1217203



Specifications

Specifications

Platinum(II) bromide
(metals basis)
MFCD00011180
KGRJUMGAEQQVFK-UHFFFAOYSA-L
dibromoplatinum
83486
Crystalline
Pt 54.5% min.
1g
13455-12-4
Br2Pt
platinum ii bromide, platinum bromide, platinum bromide ptbr2, platinum dibromide, ptbr2, acmc-1c2hr, platinum ii bromide, platinum bromide, ptbr2
Br[Pt]Br
354.892
354.91
99.9%
250°C (decomposition)

Chemical Identifiers

13455-12-4
354.892
KGRJUMGAEQQVFK-UHFFFAOYSA-L
83486
Br[Pt]Br
Br2Pt
MFCD00011180
platinum ii bromide, platinum bromide, platinum bromide ptbr2, platinum dibromide, ptbr2, acmc-1c2hr, platinum ii bromide, platinum bromide, ptbr2
dibromoplatinum
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents