Learn More
Platinum(II) bromide, 99.9% (metals basis), Pt 54.5% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01217206
Description
Platinum(II) bromide used as Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum(II) bromide | |
13455-12-4 | |
5 g | |
Br2Pt | |
platinum ii bromide, platinum bromide, platinum bromide ptbr2, platinum dibromide, ptbr2, acmc-1c2hr, platinum bromide, ptbr2 | |
KGRJUMGAEQQVFK-UHFFFAOYSA-L | |
dibromoplatinum | |
83486 | |
99.9% |
250°C (decomposition) | |
Crystalline | |
(metals basis) | |
MFCD00011180 | |
Soluble in water. | |
[Br-].[Br-].[Pt++] | |
354.89 | |
354.91 | |
Pt 54.5% min. |
Chemical Identifiers
13455-12-4 | |
354.89 | |
KGRJUMGAEQQVFK-UHFFFAOYSA-L | |
83486 | |
[Br-].[Br-].[Pt++] |
Br2Pt | |
MFCD00011180 | |
platinum ii bromide, platinum bromide, platinum bromide ptbr2, platinum dibromide, ptbr2, acmc-1c2hr, platinum bromide, ptbr2 | |
dibromoplatinum |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 236-648-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only