Please login to your online account to display your discounted pricing

Platinum(II) acetylacetonate, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC369350010

 View more versions of this product

Catalog No. AC369350010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5% (ex Pt)
Formula Weight 393.31
Physical Form Powder
Quantity 1g
Color Yellow to yellow-green
Sublimation Point 170
Molecular Formula C10H14O4Pt
CAS 15170-57-7
Health Hazard 1 Warning
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula CH3COCH=C(CH3)O!2Pt
Chemical Name or Material Platinum(II) acetylacetonate

Specifications

Chemical Name or Material Platinum(II) acetylacetonate
Assay ≥97.5% (ex Pt)
Assay Percent Range 98%
Molecular Formula C10H14O4Pt
Linear Formula CH3COCH=C(CH3)O!2Pt
Formula Weight 393.31
Accessories

Warning

SDS & Safety Documents