missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Platinum ICP Standard, 1000 ppm Pt in 3% HCl, Ricca Chemical
Supplier: Ricca Chemical Company PPT1KH500
Specifications
Platinum ICP Standard | |
Natural Poly Bottle | |
93.0,4.38,0.09 | |
Yellow | |
Pt | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
platinum | |
23939 | |
1 mL = 1 mg Pt |
7440-06-4,7647-01-0,7732-18-5 | |
500 mL | |
97.0,4.56,0.1 | |
1,000ppm Pt | |
MFCD00011179 | |
[Pt] | |
195.08 | |
CHEBI:33400 | |
Liquid |
Chemical Identifiers
7440-06-4 | |
195.08 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
CHEBI:33400 | |
[Pt] |
Pt | |
MFCD00011179 | |
23939 | |
platinum |