missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum gauze, Unimesh N7433, expanded metal mesh, 0.34mm (0.013in) thick, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 046966FF

 View more versions of this product

Catalog No. AA46966FF



Description

Description

Platinum gauze is used in electronics industry for electrical contacts which might be subjected to high temperatures. It also acts as a catalyst in the production of nitric acid. It plays an essential role in the cracking processes in oil refineries. It can be coated on electrode surface by electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Platinum gauze, Unimesh N7433
7440-06-4
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
0.34mm Mesh
≈1.0g/25x25mm
Expanded metal mesh, 0.34mm (0.013 in.) thick
Pt
25 x 25 mm
Soluble in hot aqua regia.
[Pt]
195.08
CHEBI:33400
Odorless

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

RTECSNumber : TP2160000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only