missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum gauze, Unimesh N7433, expanded metal mesh, 0.34mm (0.013 in.) thick

Manufacturer:  Alfa Aesar™ 46966FI

 View more versions of this product

Catalog No. AA46966FI



Specifications

Specifications

Platinum gauze, Unimesh N7433
50 x 50mm
7440-06-4
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
[Pt]
195.08
CHEBI:33400
≈1.0g/25x25mm
Expanded metal mesh, 0.34mm (0.013 in.) thick
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
Odorless
platinum
23939
Mesh

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

RTECSNumber : TP2160000

TSCA : TSCA

SDS Documents