missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum gauze, 99.9% (metals basis), Alfa Aesar™

Form: approx. 0.47g/25x25mm; Wire cloth; Open area: 62.7%; Width of opening: 0.015 in.

Manufacturer:  Alfa Aesar 10282FI

 View more versions of this product

Catalog No. AA10282FI



Specifications

Specifications

Platinum
7440-06-4
99.9%
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400
White
50 x 50mm
Silvery white guage
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
Odorless
23939
Gauze

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

Documents