missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum gauze, 99.9% (metals basis), Alfa Aesar™

Form: approx. 0.47g/25x25mm; Wire cloth; Open area: 62.7%; Width of opening: 0.015 in.

Manufacturer:  Thermo Fisher Scientific 10282FF

 View more versions of this product

Catalog No. AA10282FF



Specifications

Specifications

Platinum
7440-06-4
21.45g/cm3 at 20°C
Pt
black
[Pt]
Odorless
23939
Gauze
25 x 25mm
Silvery white guage
99.9%
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400
White

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

Documents