missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum foil, 99.99% (metals basis), Alfa Aesar™, Premion™

Form: approx. 1.34g/25x25mm

Manufacturer:  Alfa Aesar 11510FF

 View more versions of this product

Catalog No. AA11510FF



Specifications

Specifications

Platinum
7440-06-4
21.45g/cm3 at 20°C
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
Odorless
23939
Foil, 0.025mm (0.001 in.) thick
25 x 25mm
Silvery white foil
99.99% (Metals basis)
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400
White

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

Documents