Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum foil, 0.25mm (0.01 in.) thick, Premion™, 99.997% (metals basis)

Chemical Identifiers

Chemical Name or Material Platinum foil
CAS 7440-06-4
Name Note 0.25mm (0.01 in.) thick
Purity Grade Premion™
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA12058FF

  SDS
alfa aesar™
12058FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Odor Odorless
Assay 99.997%
Assay Percent Notes (metals basis)
Purity Grade Premion™
Quantity 25x25mm
Physical Form Foil
Name Note 0.25mm (0.01 in.) thick
CAS 7440-06-4
Chemical Name or Material Platinum foil
TSCA TSCA
Weight ≈3.35g/25x25mm
MDL Number MFCD00011179