missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum foil, 0.25mm (0.01in) thick, Premion™, 99.995% (metals basis), Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 012058FF

Catalog No. AA12058FF



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Platinum foil
7440-06-4
Pt
25 x 25 mm
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
99.997%
Odorless
Foil
0.25mm (0.01 in.) thick
(metals basis)
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
Premion™
≈3.35g/25x25mm

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

RTECSNumber : TP2160000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only