Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum foil, 0.25mm (0.01 in.) thick, 99.95% (metals basis)

Chemical Identifiers

Chemical Name or Material Platinum foil
CAS 7440-06-4
Name Note 0.25mm (0.01 in.) thick
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38378FF

  SDS
alfa aesar™
38378FF
25x25mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum foil
TSCA TSCA
Name Note 0.25mm (0.01 in.) thick
Physical Form Foil
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD00011179
CAS 7440-06-4
Odor Odorless
Weight ≈3.4g/25x25mm
Quantity 25x25mm