Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum foil, 0.25mm (0.01 in.) thick, 99.9% (metals basis)

$1,461.55 - $1,461.55

Chemical Identifiers

Chemical Name or Material Platinum foil
CAS 7440-06-4
Name Note 0.25mm (0.01 in.) thick
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA00861FF

  SDS
alfa aesar™
00861FF
25x25mm Each for $1,461.55
Description & Specifications

Specifications

Quantity 25x25mm
TSCA TSCA
Odor Odorless
Weight ≈3.4g/25x25mm
MDL Number MFCD00011179
Assay 99.9%
CAS 7440-06-4
Physical Form Foil
Assay Percent Notes (metals basis)
Chemical Name or Material Platinum foil
Name Note 0.25mm (0.01 in.) thick