Please login to your online account to display your discounted pricing

Platinum Cobalt Color Standard Solution (APHA), Fisher Chemical


Quantity 1L
Packaging Glass Bottle

Manufacturer: fisher scientific company  FLSP120-1

 View more versions of this product

Catalog No. SP120-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 100°C
Melting Point 0°C
Quantity 1L
UN Number 1789
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
pH 1.1
Color Yellow
Density 1.0g/cm³
Absorbance 0.055 to 0.065 AU at 510nm
CAS 7791-13-1
MDL Number MFCD00214189
CAS 7732-18-5
CAS 16921-30-5
CAS 7647-01-0
ChemAlert Storage Symbol White
Packaging Glass Bottle
Identification Pass Test
Chemical Name or Material Platinum Cobalt Color Standard Solution

Specifications

Chemical Name or Material Platinum Cobalt Color Standard Solution
CAS 7647-01-0
CAS 7791-13-1
CAS 16921-30-5
CAS 7732-18-5
MDL Number MFCD00214189
Accessories
 

SDS & Safety Documents