missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum black Powder, Alfa Aesar™

Form: S.A. 10m2/g

Manufacturer:  Alfa Aesar 1207606

 View more versions of this product

Catalog No. AA1207606



Specifications

Specifications

Platinum black
7440-06-4
Pt
black
[Pt]
Odorless
23939
Powder
5g
Dark gray
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400
Gray

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

H228 : Flammable solid

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-116-1

Documents