Please login to your online account to display your discounted pricing

Platinum black, Alfa Aesar™

Form: S.A. nominally 27m2/g; HiSPEC™ 1000

Chemical Identifiers

Chemical Name or Material Platinum black
CAS 7440-06-4
Physical Form Powder
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1207677

  SDS
alfa aesar
1207677
0.1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 0.1g
Color Dark gray
DOT Information DOT Class: 4.1, Packaging Group: II
EINECS Number 231-116-1
Density 21.45g/cm3 at 20°C
Physical Form Powder
CAS 7440-06-4
Health Hazard 1 H228: Flammable solid.
Odor Odorless
Appearance Dark gray powder
MDL Number MFCD00011179
Chemical Name or Material Platinum black
Safety and Handling

H228: Flammable solid.