missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum black, HiSPEC 1000, Alfa Aesar™

Manufacturer:  Alfa Aesar™ 1275503

 View more versions of this product

Catalog No. AA1275503



Specifications

Specifications

Platinum black
7440-06-4
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
Odorless
23939
Powder
1g
Dark gray powder
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
195.08
CHEBI:33400

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

H228: Flammable solid.

EINECSNumber : 231-116-1

Documents