missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Platinum on activated carbon, 5% Pt, not reduced, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 293660500
Specifications
Platinum on activated carbon | |
50g | |
7440-06-4 | |
Pt | |
01,890; 02,332 | |
black, iv ion, metal, platin, platin german, platine, platino, platinum, elemental, platinum, metal, sponge | |
[Pt] | |
195.08 | |
23939 | |
195.08 | |
5% (Pt) | |
1769.0°C to 1773.0°C |
21.45 | |
5% Pt | |
21.4500g/mL | |
MFCD00011179 | |
15, 7644 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
platinum | |
Glass bottle | |
CHEBI:33400 | |
Powder | |
Black | |
3827.0°C |
Chemical Identifiers
7440-06-4 | |
195.08 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
23939 | |
platinum |
Pt | |
MFCD00011179 | |
black, iv ion, metal, platin, platin german, platine, platino, platinum, elemental, platinum, metal, sponge | |
CHEBI:33400 | |
[Pt] |
Safety and Handling
EINECSNumber : 231-116-1
RTECSNumber : TP2160000
TSCA : TSCA