missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum on activated carbon, 5% Pt, not reduced, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 293660100

 View more versions of this product

Catalog No. AC293660100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Platinum on activated carbon
10g
7440-06-4
Pt
01,890; 02,332
black, iv ion, metal, platin, platin german, platine, platino, platinum, elemental, platinum, metal, sponge
[Pt]
195.08
23939
195.08
5% (Pt)
1769.0°C to 1773.0°C
21.45
5% Pt
21.4500g/mL
MFCD00011179
15, 7644
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
Glass bottle
CHEBI:33400
Powder
Black
3827.0°C

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, iv ion, metal, platin, platin german, platine, platino, platinum, elemental, platinum, metal, sponge
CHEBI:33400
[Pt]
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

RTECSNumber : TP2160000

TSCA : TSCA

SDS Documents
Product Certifications