Please login to your online account to display your discounted pricing

Platinum on activated carbon, 5% Pt, not reduced, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Platinum on activated carbon
Name Note 5% Pt
CAS 2023600
Assay 5% (Pt)
Molecular Formula Pt
Formula Weight 195.08
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC293660100

  SDS
thermo fisher scientific
293660100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC293660500

  SDS
thermo fisher scientific
293660500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 195.08
Quantity 10g
Color Black
Fieser 01,890; 02,332
Merck Index 15, 7644
Assay 5% (Pt)
RTECS Number TP2160000
EINECS Number 231-116-1
TSCA TSCA
Density 21.45g/mL
CAS 2023600
Name Note 5% Pt
Molecular Formula Pt
Chemical Name or Material Platinum on activated carbon
MDL Number MFCD00011179
Packaging Glass bottle