Please login to your online account to display your discounted pricing

Platinum on activated carbon, 5% Pt, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  195230100

 View more versions of this product

Catalog No. AC195230100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 195.08
Quantity 10g
Color Black
Assay 5% (Pt)
Particle Size Particle size distribution: <7 Micron: 10%, <28 Micron: 50%, <85 Micron: 90%
Fieser 01,982; 08,405; 04,393; 04,393; 04,393; 04,393
Chemical Name or Material Platinum on activated carbon
Molecular Formula Pt
CAS 7440-06-4
Name Note 5% Pt
MDL Number MFCD00011179
Surface Area 900m2/g
Packaging Glass bottle

Specifications

Chemical Name or Material Platinum on activated carbon
Name Note 5% Pt
CAS 7440-06-4
Assay 5% (Pt)
Molecular Formula Pt
Formula Weight 195.08
MDL Number MFCD00011179