Learn More
Platinum on activated carbon, 10% Pt, ca .50% moisture, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 195240100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Black | |
7440-06-4 | |
MFCD00011179 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
[Pt] | |
195.08 | |
CHEBI:33400 | |
10% (Pt) | |
Powder |
Platinum on activated carbon | |
Pt | |
10 g | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
platinum | |
23939 | |
195.08 | |
Glass bottle |
Chemical Identifiers
7440-06-4 | |
195.08 | |
BASFCYQUMIYNBI-UHFFFAOYSA-N | |
23939 | |
platinum |
Pt | |
MFCD00011179 | |
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
CHEBI:33400 | |
[Pt] |
Safety and Handling
GHS H Statement:
Self-heating in large quantities; may catch fire.
GHS P Statement:
Keep cool. Protect from sunlight.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only