Please login to your online account to display your discounted pricing

Platinum on activated carbon, 1% Pt, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Platinum on activated carbon
Name Note 1% Pt
CAS 7440-06-4
Assay 1% (Pt)
Molecular Formula Pt
Formula Weight 195.08
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC195220250

  SDS
thermo fisher scientific
195220250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC195221000

  SDS
thermo fisher scientific
195221000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.08
Physical Form Powder
Quantity 25g
Color Black
Assay 1% (Pt)
Particle Size Typical particle size distribution: D50: 28 Micron, <8 Micron: 14%, <16 Micron: 29%, <32 Micron: 55%, <64 Micron: 84%, <128 Micron: 99%
Chemical Name or Material Platinum on activated carbon
Name Note 1% Pt
CAS 7440-06-4
MDL Number MFCD00011179
Molecular Formula Pt
Packaging Glass bottle