Please login to your online account to display your discounted pricing

Platinum on activated carbon, 1% Pt, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  195221000

 View more versions of this product

Catalog No. AC195221000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Platinum on activated carbon
Quantity 100g
Name Note 1% Pt
CAS 7440-06-4
Physical Form Powder
Assay 1% (Pt)
Molecular Formula Pt
Formula Weight 195.08
MDL Number MFCD00011179
Packaging Glass bottle
Particle Size Typical particle size distribution: D50: 28 Micron, <8 Micron: 14%, <16 Micron: 29%, <32 Micron: 55%, <64 Micron: 84%, <128 Micron: 99%
Color Black

Specifications

Chemical Name or Material Platinum on activated carbon
Name Note 1% Pt
CAS 7440-06-4
Assay 1% (Pt)
Molecular Formula Pt
Formula Weight 195.08
MDL Number MFCD00011179