missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Platinum, AAS standard solution, Pt 1000μg/mL, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 88086AE

 View more versions of this product

Catalog No. AA88086AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Platinum
Pt 1000μg/mL
MFCD00011179
Characteristic
100mL
7440-6-6
Pt in 20% HCl
Liquid
Approx. 1g/cm3 at 20°C

Chemical Identifiers

7440-6-6
Pt in 20% HCl
MFCD00011179
Safety and Handling

Safety and Handling

H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 231-595-7

SDS Documents