Please login to your online account to display your discounted pricing

Platinum, AAS standard solution, Pt 1000μg/mL, Alfa Aesar™, Specpure™

Chemical Identifiers

Chemical Name or Material Platinum
Physical Form Liquid
Assay Percent Range Pt 1000μg/mL
Molecular Formula Pt in 20% HCl
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA88086AE

 
alfa aesar
88086AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA88086AP

 
alfa aesar
88086AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 100mL
DOT Information DOT Class: 8, Packaging Group: II
Chemical Name or Material Platinum
EINECS Number 231-595-7
Physical Form Liquid
Assay Percent Range Pt 1000μg/mL
Molecular Formula Pt in 20% HCl
MDL Number MFCD00011179
Health Hazard 1 H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.
Odor Characteristic
Density Approx. 1g/cm3 at 20°C
Safety and Handling

H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.