Please login to your online account to display your discounted pricing

Platinum, 99.9%, (trace metal basis), powder, 0.27-0.47 microns, ACROS Organics™

$155.41 - $155.41

Chemical Identifiers

Chemical Name or Material Platinum
Name Note 0.27-0.47 micron, 99.9%
CAS 7440-06-4
Purity Grade Trace Metal Basis
Assay Percent Range 99.9%
Molecular Formula Pt
Linear Formula Pt
Formula Weight 195.08
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC318855000

  SDS
thermo fisher scientific
318855000
Glass bottle 500mg Each for $155.41
Description & Specifications

Specifications

Purity Grade Trace Metal Basis
Physical Form Powder
Formula Weight 195.08
Quantity 500mg
Color Gray to black
Fieser 01,890; 02,332
Merck Index 15, 7644
EINECS Number 231-116-1
TSCA TSCA
Density 21.45
RTECS Number TP2160000
Chemical Name or Material Platinum
Name Note 0.27-0.47 micron, 99.9%
Assay Percent Range 99.9%
Molecular Formula Pt
Linear Formula Pt
MDL Number MFCD00011179
CAS 7440-06-4
Packaging Glass bottle
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Flammable solid.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Total Trace Metal Impurities 0.1% max.

For ultra trace analysis

Safety and Handling

Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.