Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 5% on graphite, Type 286, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Platinum
Name Note 5% on graphite
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4436506

 
alfa aesar™
4436506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4436514

 
alfa aesar™
4436514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material Platinum
Name Note 5% on graphite
Physical Form Powder
MDL Number MFCD00011179
Health Hazard 1 H228-H315-H319-H335 : Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Appearance Powder
Safety and Handling

H228-H315-H319-H335 : Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.