Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 5% on carbon paste, B501018-5

Chemical Identifiers

Chemical Name or Material Platinum
CAS 7440-06-4
Name Note 5% on carbon paste, B501018-5
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3631006

  SDS
alfa aesar™
H3631006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3631014

  SDS
alfa aesar™
H3631014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3631022

  SDS
alfa aesar™
H3631022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum
Name Note 5% on carbon paste, B501018-5
CAS 7440-06-4
TSCA TSCA
Physical Form Solid
MDL Number MFCD00011179
Odor Odorless
Color Dark gray
Quantity 5g