missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 5% on carbon, dry

Manufacturer:  Alfa Aesar™ A1118614

 View more versions of this product

Catalog No. AAA1118614


 

Description

Description

Applications
Platinum is a catalyst used for hydrgenation of alkenes, alkynes, nitro groups; also used for hydrogenolysis of cyclopropanes, catalyzes oxidation of alcohols.

Solubility
Soluble in boiling aqua regia (reacts), and molten alkali cyanides (reacts). Insoluble in water, mineral acids, and organic acids.

Notes
Do not store together with acids. Incompatible with Strong acids, strong oxidizing agents & organics Solvent vapors. Store under inert gas.
Specifications

Specifications

Platinum
7440-06-4
MFCD00011179
14,7529
[Pt]
195.08
CHEBI:33400
5% on carbon, Dry
Pt
UN1325
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
25g

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
CHEBI:33400
Pt
MFCD00011179
23939
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : TP2160000

TSCA : TSCA

SDS Documents