missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 5% on alumina powder, B301099-5

Manufacturer:  Alfa Aesar™ H3623406

 View more versions of this product

Catalog No. AAH3623406


 

Specifications

Specifications

Platinum
7440-06-4
MFCD00011179
[Pt]
195.08
CHEBI:33400
Odorless
5g
5% on alumina powder, B301099-5
Pt
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Powder Pellets or Spheres
Gray

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
CHEBI:33400
Pt
MFCD00011179
23939
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents