Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 5% on alumina powder, B301099-5

Chemical Identifiers

Chemical Name or Material Platinum
CAS 7440-06-4
Name Note 5% on alumina powder, B301099-5
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3623406

  SDS
alfa aesar™
H3623406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3623414

  SDS
alfa aesar™
H3623414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3623422

  SDS
alfa aesar™
H3623422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum
TSCA TSCA
Odor Odorless
Name Note 5% on alumina powder, B301099-5
Physical Form Powder Pellets or Spheres
MDL Number MFCD00011179
CAS 7440-06-4
Color Gray
Quantity 5g