missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 5% on activated carbon powder, sulfided, 0.5% S (as sulfide)

Manufacturer:  Alfa Aesar™ 4390518

 View more versions of this product

Catalog No. AA4390518

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Platinum
5% on activated carbon powder, Sulfided, 0.5% S (as sulfide)
MFCD00011179
Powder
50g
7440-6-6
UN1325

Chemical Identifiers

7440-6-6
MFCD00011179
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents