Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 5% on activated carbon paste, Type

Chemical Identifiers

Chemical Name or Material Platinum
CAS 7440-06-4
Name Note 5% on activated carbon paste, Type 128M
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3764106

  SDS
alfa aesar™
H3764106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3764122

  SDS
alfa aesar™
H3764122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3764114

  SDS
alfa aesar™
H3764114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum
Name Note 5% on activated carbon paste, Type 128M
CAS 7440-06-4
TSCA TSCA
Physical Form Solid
MDL Number MFCD00011179
Odor Odorless
Color Black
Quantity 5g