Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum, 5% on activated carbon paste, Type 18

Chemical Identifiers

Chemical Name or Material Platinum
CAS 7440-06-4
Name Note 5% on activated carbon paste, Type 18
MDL Number MFCD00011179
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3696814

  SDS
alfa aesar™
H3696814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3696806

  SDS
alfa aesar™
H3696806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3696822

  SDS
alfa aesar™
H3696822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Platinum
Name Note 5% on activated carbon paste, Type 18
CAS 7440-06-4
TSCA TSCA
Physical Form Solid
MDL Number MFCD00011179
Odor Odorless
Color Dark gray
Quantity 25g