missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Platinum 30 wt% Rhodium wire, 0.5mm (0.02 in.) dia., ISA Type B Standard Grade Thermocouple

For ISA Type B Standard Grade Thermocouple

Manufacturer:  Alfa Aesar™ 12576BY

 View more versions of this product

Catalog No. AA12576BY

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Platinum 30 wt% Rhodium wire
MFCD00198186
Wire
100cm
0.5mm (0.02 in.) dia.
Pt:Rh; 70:30 wt%
Odorless
≈0.95g/25cm
Documents